Methyl coumalate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-oxo-2H-pyran-5-carboxylate

A381504-250MG A381504-10G
103826-700EA 7 USD
103826-700 103837-734 77594-144 77686-065
Methyl coumalate 98%
Methyl coumalate

Precaución: Inert atmosphere, room temperature

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 265.9 °C (760 mm Hg)
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00010120
CAS Number: 6018-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR