Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 1,2-dihydro-2-oxopyridine-3-carboxylate, 1,2-Dihydro-3-(methoxycarbonyl)-2-oxopyridine

A153988-250MG A153988-10G
103804-588EA 22 USD
103804-588 103842-768 77010-928 77683-293
Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate 97%, Ambeed.Inc
Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate

Precaución: For research use only.

Formula: C₇H₇NO₃
MW: 153.137 g/mol
Boiling Pt: 367.6 °C (760 mm Hg)
Melting Pt: 115…118 °C
Flash Pt: 176.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00661282
CAS Number: 10128-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR