Methyl nitroacetate 96%

Supplier: Ambeed
Warning

Synonyms: Nitroacetic methyl ester, Methyl 2-nitroacetate

A104731-1G A104731-5G A104731-25G A104731-500G A104731-100G
103796-768EA 6 USD
103796-768 103822-492 77309-350 77309-352 77610-818 77684-950 77901-486
Methyl nitroacetate 96%
Methyl nitroacetate

Precaución: for Research Use Only

Formula: C₃H₅NO₄
Boiling Pt: 196.5 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00075480
CAS Number: 2483-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR