Methyl-2-naphthoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-naphthoate

A134895-100G A134895-10G A134895-250MG A134895-1G A134895-5G
103802-070EA 6 USD
103802-070 76798-014 76798-016 76798-018 77314-838 77314-840
Methyl-2-naphthoate 97%
Methyl 2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 290 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 145.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00017345
CAS Number: 2459-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR