Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Piroxicam Related Compound J, Methyl 2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide
A216100-10G
A216100-250MG
A216100-1G
A216100-5G
A216100-100G
103818-536EA
25
USD
103818-536
103849-248
76975-700
76975-702
77087-820
77087-822
77881-530
Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide 97%
Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide
Precaución: for Research Use Only
Formula:
C₁₁H₁₁NO₅S Boiling Pt: 439.9 °C (760 mm Hg) Melting Pt: 163…165 °C Flash Pt: 219.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00071766 CAS Number: 35511-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...