Methyl-2-methyl-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-3-nitrobenzoate

A235851-25G A235851-100G A235851-10G A235851-500G
103850-808EA 5 USD
103850-808 77089-826 77275-894 77275-896 77275-898
Methyl-2-methyl-3-nitrobenzoate 98%
Methyl 2-methyl-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Boiling Pt: 286.6 °C (760 mm Hg)
Melting Pt: 62…65 °C
Flash Pt: 128.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00082564
CAS Number: 59382-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR