Methyl-2,2-dimethylphenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-2-phenylpropanoate

A215824-1G A215824-25G
103818-516EA 6 USD
103818-516 103849-224 77087-776 77303-418
Methyl-2,2-dimethylphenylacetate 98%
Methyl-2,2-dimethylphenylacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01846451
CAS Number: 57625-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR