Methyl 2-methyl-1H-indole-3-carboxylate 98%

Supplier: Ambeed
Warning

A161658-100MG A161658-250MG A161658-25G A161658-1G A161658-5G A161658-10G
103805-512EA 23 USD
103805-512 103843-750 77011-222 77011-224 77011-226 77300-544 77300-546 77611-572
Methyl 2-methyl-1H-indole-3-carboxylate 98%
Methyl 2-methyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Boiling Pt: 331.7±22.0 °C (760 mm Hg)
Melting Pt: 165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00963961
CAS Number: 65417-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR