Methyl 2-mercapto-1H-benzo[d]imidazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A481800-100MG
A481800-250MG
103828-130EA
42
USD
103828-130
103839-204
77143-810
77143-812
Methyl 2-mercapto-1H-benzo[d]imidazole-4-carboxylate 97%
Methyl 2-mercapto-1H-benzo[d]imidazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂S MW: 208.24 g/mol Boiling Pt: 350.3±44.0 °C (760 mm Hg) Flash Pt: 165.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD24600834 CAS Number: 92807-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...