Methyl glycolate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hydroxyacetic Acid Methyl Ester, Methyl 2-hydroxyacetate, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate
A152116-1000G
A152116-100G
A152116-10G
A152116-1G
A152116-500G
A152116-25G
A152116-5G
103842-508EA
7
USD
103842-508
77110-350
77110-352
77110-354
77110-356
77110-358
77318-118
77318-120
Methyl glycolate 98%
Methyl glycolate
Precaución: for Research Use Only
Formula:
C₃H₆O₃ MW: 90.08 g/mol Boiling Pt: 149…151 °C (760 mm Hg) Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004667 CAS Number: 96-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...