Methyl 2-hydroxy-6-methylbenzoate 95%

Supplier: Ambeed
Warning

A139237-100MG A139237-250MG A139237-1G A139237-5G
103802-714EA 80 USD
103802-714 76811-680 76811-682 77299-056 77299-058
Methyl 2-hydroxy-6-methylbenzoate 95%
Methyl 2-hydroxy-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
Boiling Pt: 230.2±20.0 °C (760 mm Hg)
Melting Pt: 31…32 °C
Flash Pt: 91.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06797968
CAS Number: 33528-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR