Methyl-4-methylsalicylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-hydroxy-4-methylbenzoate
A145940-100G
A145940-10G
A145940-5G
103803-606EA
6
USD
103803-606
103841-690
76689-180
76689-182
77316-994
Methyl-4-methylsalicylate 98%
Methyl 4-methylsalicylate
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 241.2 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00020130 CAS Number: 4670-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...