Methyl-4-iodsalicylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-hydroxy-4-iodobenzoate
A141637-100G
A141637-10G
A141637-1G
A141637-25G
A141637-5G
103803-032EA
5
USD
103803-032
76782-948
76782-950
77316-198
77316-200
77316-202
Methyl-4-iodsalicylate 97%
Methyl 4-iodsalicylate
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ Boiling Pt: 309.7 °C (760 mm Hg) Melting Pt: 69…73 °C Flash Pt: 141.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797864 CAS Number: 18179-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...