Methyl-2-hydroxy-3-phenylpropanoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-3-phenylpropanoate

A320937-1G A320937-25G A320937-10G A320937-250MG A320937-5G
103825-258EA 6 USD
103825-258 103836-190 76839-076 76839-078 77263-402 77263-404 77306-230
Methyl-2-hydroxy-3-phenylpropanoate 97%
Methyl-2-hydroxy-3-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06654233
CAS Number: 13674-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR