Methyl-2-formylbenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-formylbenzoate
A105884-100G
A105884-10G
A105884-25G
A105884-1G
A105884-5G
103796-960EA
12
USD
103796-960
103822-706
77257-434
77257-436
77257-438
77283-406
77309-570
Methyl-2-formylbenzoate 98%
Methyl-2-formylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈O₃ MW: 164.1580 g/mol Boiling Pt: 280 °C (760 mm Hg) Flash Pt: 122.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204231 CAS Number: 4122-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...