Methyl-2-fluoro-6-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-6-methylbenzoate

A211991-25G A211991-5G
103818-152EA 17 USD
103818-152 103848-848 77087-282 77087-284
Methyl-2-fluoro-6-methylbenzoate 97%
Methyl 2-fluoro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.1700 g/mol
Boiling Pt: 215.8±28.0 °C (760 mm Hg)
Flash Pt: 82.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11226310
CAS Number: 197516-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR