Methyl-2-fluoro-5-nitrobenzenecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-5-nitrobenzenecarboxylate, Methyl 2-fluoro-5-nitrobenzoate

A101088-100G A101088-10G A101088-250MG A101088-25G A101088-5G A101088-1G
103795-800EA 6 USD
103795-800 103821-490 77008-310 77008-312 77008-314 77008-316 77282-658 77296-986
Methyl-2-fluoro-5-nitrobenzenecarboxylate 97%
Methyl-2-fluoro-5-nitrobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Boiling Pt: 302.8 °C (760 mm Hg)
Melting Pt: 49…51 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD06203689
CAS Number: 2965-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR