Methyl-2-fluoro-4-iodonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-4-iodonicotinate

A100958-100G A100958-100MG A100958-250MG A100958-1G A100958-5G
103795-724EA 11 USD
103795-724 103821-414 77107-180 77107-182 77107-184 77296-928 77296-930
Methyl-2-fluoro-4-iodonicotinate 97%
Methyl-2-fluoro-4-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₅FINO₂
MW: 281.0200 g/mol
Boiling Pt: 287 °C (760 mm Hg)
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972401
CAS Number: 884494-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR