Methyl-2-fluoro-4-formylbenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-fluoro-4-formylbenzoate
A259317-10G
A259317-250MG
A259317-1G
A259317-25G
A259317-5G
103812-430EA
25
USD
103812-430
103852-310
76874-824
76874-826
77091-852
77091-854
77091-856
Methyl-2-fluoro-4-formylbenzoate 97%
Methyl-2-fluoro-4-formylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₇FO₃ MW: 182.15 g/mol Melting Pt: 70…74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20231454 CAS Number: 85070-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...