Methyl 2-Fluoro-2-methylpropionate 97%

Supplier: Ambeed
Danger

A854074-10G A854074-5G A854074-1G
103841-246EA 14 USD
103841-246 76728-866 76845-502 77308-456
Methyl 2-Fluoro-2-methylpropionate 97%
Methyl 2-Fluoro-2-methylpropionate

Precaución: for Research Use Only

Formula: C₅H₉FO₂
Boiling Pt: 112.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03265444
CAS Number: 338-76-1
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR