Methyl 2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A157288-250MG
A157288-25G
A157288-5G
103804-984EA
9
USD
103804-984
103843-184
76823-986
77319-000
77319-002
Methyl 2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 98%
Methyl 2-ethoxy-1H-benzo[d]imidazole-7-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O₃ MW: 220.23 g/mol Boiling Pt: 373.1 °C (760 mm Hg) Melting Pt: 130…135 °C Flash Pt: 179.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD14636510 CAS Number: 150058-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...