Methyl-2-cyano-2-methylpropanoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-cyano-2-methylpropanoate, Methyl 2-cyano-2,2-dimethylacetate

A109965-10G A109965-1G A109965-5G
103797-596EA 5 USD
103797-596 103823-404 77245-650 77310-210 77310-212
Methyl-2-cyano-2-methylpropanoate 97%
Methyl-2-cyano-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.14 g/mol
Boiling Pt: 186.5 °C (760 mm Hg)
Flash Pt: 74.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03428328
CAS Number: 72291-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR