Methyl-2-chloropyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloropyrimidine-5-carboxylate

A177135-100G A177135-10G A177135-250MG A177135-1G A177135-25G A177135-5G
103807-214EA 8 USD
103807-214 103845-540 76799-552 76799-554 76799-556 77082-920 77082-922 77082-924
Methyl-2-chloropyrimidine-5-carboxylate 98%
Methyl-2-chloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.5691 g/mol
Boiling Pt: 242.8 °C (760 mm Hg)
Melting Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690330
CAS Number: 287714-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR