Methyl-2-chloropyrimidine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A279800-250MG
A279800-100G
103813-486EA
34
USD
103813-486
103853-430
77668-238
77872-346
Methyl-2-chloropyrimidine-4-carboxylate 97%
Methyl-2-chloropyrimidine-4-carboxylate
Precaución: Inert atmosphere, 2…8 °C.
Formula:
C₆H₅ClN₂O₂ Melting Pt: 97…98 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11841080 CAS Number: 149849-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...