(2-Chlorophenyl)acetic acid methyl ester 98%

Supplier: Ambeed
Warning

A323611-100G A323611-10G A323611-500G A323611-1000G A323611-25G
103810-126EA 144 USD
103810-126 77277-738 77277-740 77277-742 77306-306 77306-308
(2-Chlorophenyl)acetic acid methyl ester 98%
(2-Chlorophenyl)acetic acid methyl ester

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOCH₃
Boiling Pt: 240.3 °C (760 mm Hg)
Flash Pt: 108.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00137445
CAS Number: 57486-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR