Methyl 2-chloro-1,3-oxazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(methoxycarbonyl)-1,3-oxazole, Methyl 2-chlorooxazole-5-carboxylate

A512609-250MG A512609-1G
103828-412EA 114 USD
103828-412 103839-512 76818-806 77307-664
Methyl 2-chloro-1,3-oxazole-5-carboxylate 97%
Methyl 2-chloro-1,3-oxazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₄ClNO₃
MW: 161.543 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09265527
CAS Number: 934236-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR