Methyl-2-chloroisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate

A128003-100G A128003-10G A128003-1G A128003-500G A128003-25G
77161-700EA 71.07 USD
77161-700 77161-702 77161-704 77161-706 77313-342
Methyl-2-chloroisonicotinate 98%
Methyl 2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.5810 g/mol
Melting Pt: 32…36 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01765409
CAS Number: 58481-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR