2-Chloro-6-methylnicotinic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-6-methylnicotinate

A162017-1G A162017-25G A162017-100G A162017-10G A162017-250MG
103805-562EA 21 USD
103805-562 103843-808 77319-834 77319-836 77871-814 77871-815 77871-816
2-Chloro-6-methylnicotinic acid methyl ester 98%
2-Chloro-6-methylnicotinic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
Boiling Pt: 252.5 °C (760 mm Hg)
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11845763
CAS Number: 53277-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR