Methyl-2-chloro-6-methoxynicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-6-methoxynicatinate, Methyl 2-chloro-6-methoxynicotinate

A396530-100MG A396530-250MG A396530-1G A396530-25G A396530-5G
103826-982EA 16 USD
103826-982 103838-022 76803-910 76803-912 77307-042 77307-044 77307-046
Methyl-2-chloro-6-methoxynicotinate 97%
Methyl-2-chloro-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.61
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD08275100
CAS Number: 95652-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR