Methyl 2-chloro-6-fluorobenzoate 97%

Supplier: Ambeed
Warning

A145368-10G A145368-250MG A145368-1G A145368-25G A145368-5G
103803-526EA 19 USD
103803-526 103841-612 76823-622 76823-624 77316-896 77316-898 77316-900
Methyl 2-chloro-6-fluorobenzoate 97%
Methyl 2-chloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02245077
CAS Number: 151360-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR