Methyl-2-chloro-5-nitronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-5-nitronicotinate

A240870-100MG A240870-250MG
103811-368EA 21 USD
103811-368 103851-174 77098-472 77098-474
Methyl-2-chloro-5-nitronicotinate 97%
Methyl-2-chloro-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Boiling Pt: 321.5 °C (760 mm Hg)
Flash Pt: 148.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368888
CAS Number: 190271-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR