Methyl 2-chloro-5-nitroisonicotinate 97%

Supplier: Ambeed
Warning

A869420-10G A869420-25G A869420-1G A869420-5G
103841-280EA 69 USD
103841-280 77268-678 77268-680 77308-472 77308-474
Methyl 2-chloro-5-nitroisonicotinate 97%
Methyl 2-chloro-5-nitroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105213
CAS Number: 777899-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR