2-Chloro-4-iodonicotinic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-4-iodonicotinate, Methyl 2-chloro-4-iodopyridine-3-carboxylate

A102884-100MG A102884-250MG A102884-1G
103796-440EA 29 USD
103796-440 103822-142 77008-414 77008-416 77309-014
2-Chloro-4-iodonicotinic acid methyl ester 97%
2-Chloro-4-iodonicotinic acid methyl ester

Precaución: for Research Use Only

Formula: C₇H₅ClINO₂
MW: 297.48 g/mol
Boiling Pt: 304.4 °C (760 mm Hg)
Melting Pt: 85…86 °C
Flash Pt: 137.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039871
CAS Number: 185041-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR