Methyl 2-chloro-3-(trifluoromethyl)isonicotinate 95%

Supplier: Ambeed
Warning

A186049-100MG A186049-1G
103815-904EA 111 USD
103815-904 103846-478 77191-812 77302-012
Methyl 2-chloro-3-(trifluoromethyl)isonicotinate 95%
Methyl 2-chloro-3-(trifluoromethyl)isonicotinate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611604
CAS Number: 1227575-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR