Methyl-2-bromothiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromothiazole-5-carboxylate

A141302-100G A141302-10G A141302-1G A141302-5G
103802-974EA 8 USD
103802-974 76984-200 76984-202 77316-116 77316-118
Methyl-2-bromothiazole-5-carboxylate 97%
Methyl-2-bromothiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂S
MW: 222.05 g/mol
Boiling Pt: 263.3 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD03788565
CAS Number: 54045-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR