Methyl-2-bromothiazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-bromothiazole-4-carboxylate
A139839-100G
A139839-250MG
A139839-1G
A139839-25G
A139839-5G
A139839-10G
103802-782EA
6
USD
103802-782
76984-136
76984-138
77315-878
77315-880
77315-882
77592-654
Methyl-2-bromothiazole-4-carboxylate 97%
Methyl-2-bromothiazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₅H₄BrNO₂S MW: 222.06 g/mol Boiling Pt: 263.3 °C (760 mm Hg) Melting Pt: 118…122 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659908 CAS Number: 170235-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...