Methyl-2-bromonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromonicotinate, Methyl 2-bromopyridine-3-carboxylate

A619777-1G A619777-5G A619777-100MG A619777-250MG
103840-208EA 36 USD
103840-208 76844-382 76844-384 77266-852 77266-854
Methyl-2-bromonicotinate 97%
Methyl-2-bromonicotinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 255.3 °C (760 mm Hg)
Melting Pt: 32…37 °C
Flash Pt: 108.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07367915
CAS Number: 52718-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR