Methyl-2-(bromomethyl)benzoate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 2-bromomethylbenzoate
A305665-5G
A305665-100G
A305665-10G
A305665-1G
A305665-25G
103814-442EA
7
USD
103814-442
103835-608
76838-306
77167-032
77167-034
77305-914
77305-916
Methyl-2-(bromomethyl)benzoate 95%
Methyl 2-(bromomethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ MW: 229.0706 g/mol Boiling Pt: 293.9 °C (760 mm Hg) Melting Pt: 30…35 °C Flash Pt: 131.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03425900 CAS Number: 2417-73-4 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...