Methyl-2-bromobenzo[d]thiazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A600498-1G A600498-5G A600498-100MG A600498-10G A600498-250MG
103828-974EA 33 USD
103828-974 103840-110 76844-266 76844-268 77035-112 77872-989 77872-990
Methyl-2-bromobenzo[d]thiazole-6-carboxylate 97%
Methyl-2-bromobenzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD16660646
CAS Number: 1024583-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR