Methyl-2-bromo-6-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-bromo-6-methylbenzoate
A182378-100G
A182378-10G
A182378-1G
A182378-25G
A182378-5G
103807-704EA
9
USD
103807-704
103846-076
76784-626
76784-628
77083-614
77083-616
77083-618
Methyl-2-bromo-6-methylbenzoate 98%
Methyl 2-bromo-6-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ MW: 229.0700 g/mol Boiling Pt: 254.3 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09260938 CAS Number: 99548-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...