Methyl 2-bromo-6-iodobenzoate 95%

Supplier: Ambeed
Warning

A101215-100MG A101215-10G A101215-250MG A101215-1G
103795-862EA 27 USD
103795-862 103821-546 77093-498 77093-500 77093-502 77297-024
Methyl 2-bromo-6-iodobenzoate 95%
Methyl 2-bromo-6-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870176
CAS Number: 1261840-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR