Methyl-2-bromo-6-fluorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromo-6-fluorobenzoate, 2-Bromo-6-fluorobenzoic acid methyl ester

A101946-100G A101946-100MG A101946-10G A101946-5G A101946-1G A101946-25G
103796-214EA 9 USD
103796-214 76821-268 76821-270 76821-272 76821-274 77308-798 77884-925
Methyl-2-bromo-6-fluorobenzoate 95%
Methyl-2-bromo-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000922
CAS Number: 820236-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR