Methyl-2-bromo-5-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromobenzoic acid methyl ester, Methyl 2-bromo-5-nitrobenzoate
A173703-25G
A173703-5G
A173703-100G
A173703-10G
A173703-1G
A173703-500G
103845-160EA
8
USD
103845-160
77082-428
77082-430
77236-344
77236-346
77236-348
77615-452
Methyl-2-bromo-5-nitrobenzoate 97%
Methyl-2-bromo-5-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrNO₄ MW: 260.04 g/mol Boiling Pt: 326.7 °C (760 mm Hg) Melting Pt: 79…81 °C Flash Pt: 151.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010867 CAS Number: 6942-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...