Methyl-2-bromo-5-methylbenzoate 95%

Supplier: Ambeed
Warning

A117858-100G A117858-25G A117858-1G A117858-5G A117858-10G
103798-946EA 20 USD
103798-946 103824-838 76810-284 76810-286 77297-560 77311-620 77670-662
Methyl-2-bromo-5-methylbenzoate 95%
Methyl 2-bromo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Boiling Pt: 270.1 °C (760 mm Hg)
Flash Pt: 117.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00144764
CAS Number: 90971-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR