Methyl-2-bromo-5-methoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromo-5-methoxybenzoate, 2-Bromo-5-methoxybenzoic acid methyl ester

A375413-1G A375413-25G A375413-5G A375413-10G A375413-100G
103826-564EA 5 USD
103826-564 103837-588 76840-874 76840-876 76840-878 77278-488 77748-751
Methyl-2-bromo-5-methoxybenzoate 98%
Methyl-2-bromo-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 291.9 °C (760 mm Hg)
Flash Pt: 130.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00051594
CAS Number: 35450-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR