Methyl 2-bromo-5-formylbenzoate 95%

Supplier: Ambeed
Warning

A120639-10G A120639-250MG A120639-1G A120639-5G
103799-976EA 93 USD
103799-976 77258-094 77258-096 77312-086 77312-088
Methyl 2-bromo-5-formylbenzoate 95%
Methyl 2-bromo-5-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD26863463
CAS Number: 1043418-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR