Methyl-2-bromo-4-fluorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromo-4-fluorobenzoate

A212242-100G A212242-10G A212242-500G A212242-25G A212242-5G A212242-1G
103818-178EA 9 USD
103818-178 103848-878 76800-670 76800-672 76800-674 77087-334 77087-336 77303-236
Methyl-2-bromo-4-fluorobenzoate 97%
Methyl-2-bromo-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.04
Boiling Pt: 241.3 °C (760 mm Hg)
Flash Pt: 99.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06204026
CAS Number: 653-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR