Methyl 2-bromo-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

A100994-10G A100994-25G A100994-1G A100994-5G
103795-740EA 20 USD
103795-740 103821-432 76765-150 76765-152 77296-938 77296-940
Methyl 2-bromo-3-nitrobenzoate 97%
Methyl 2-bromo-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MW: 260.042 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Flash Pt: 135.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00087891
CAS Number: 5337-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR