Methyl-2-aminothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-aminothiazole-4-carboxylate

A204395-25G A204395-5G A204395-100G A204395-10G A204395-1G A204395-500G
103848-212EA 6 USD
103848-212 77086-384 77086-386 77287-754 77287-756 77287-758 77287-760
Methyl-2-aminothiazole-4-carboxylate 97%
Methyl 2-aminothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.1800 g/mol
Flash Pt: 134.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00622441
CAS Number: 118452-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR