Methyl-2-aminopyrimidine-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-aminopyrimidine-5-carboxylate
A174185-250MG
A174185-25G
A174185-1G
A174185-5G
103806-932EA
17
USD
103806-932
103845-230
76784-342
76784-344
77082-526
77301-326
Methyl-2-aminopyrimidine-5-carboxylate 97%
Methyl-2-aminopyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇N₃O₂ MW: 153.1400 g/mol Boiling Pt: 344.2 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08275693 CAS Number: 308348-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...